Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (76-2-11), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128238
Description
CD8 Monoclonal specifically detects CD8 in Porcine samples. It is validated for Flow Cytometry.Specifications
CD8 | |
Monoclonal | |
0.5mg/mL | |
Flow Cytometry <= 1 ug/10^6 cells | |
CD8A | |
Fresh dd miniature swine thymocytes | |
0.25 mg | |
Innate Immunity, Stem Cell Markers | |
925 | |
Pig | |
Purified |
Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
76-2-11 | |
FITC | |
CD8, CD8a molecule, LEU2, p32 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Porcine CD8a, Mr 32-34 kDa | |
Store at 4C. Do not freeze. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction