Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (7C2B), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260959
Description
CD8 Monoclonal specifically detects CD8 in Bovine, Goat, Bison, Water Buffalo, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD8 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10 - 1:1000 | |
CD8, CD8a molecule, LEU2, p32 | |
Mouse | |
Unpurified | |
Primary | |
Bovine, Goat, Bison, Water Buffalo, Sheep | |
IgG2a |
Flow Cytometry, Immunohistochemistry, Functional Assay | |
7C2B | |
Unconjugated | |
Ascites with 0.09% Sodium Azide | |
CD8A | |
Sheep mononuclear leukocyytes | |
0.1 mg | |
925 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction