Learn More
CD8 Antibody (fCD8), DyLight 680, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP128254FR
Catalog No. NBP128254FR
Only null left
Add to Cart
Description
Description
CD8 Monoclonal specifically detects CD8 in Feline samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
CD8
Monoclonal
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
CD8A
Feline CD8, Mr 70 kDa (38/31 kDa under reducing conditions).
0.1 mL
Adaptive Immunity, Cytokine Research, Immunology, Immunophenotyping, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers
925
Feline
Purified
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
fCD8
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
CD8, CD8a molecule, LEU2, p32
Mouse
Protein A or G purified
RUO
Primary
Feline CD8, Mr 70 kDa (38/31 kDa under reducing conditions).
Store at 4C in the dark.
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.