Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (fCD8), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128254IR
Description
CD8 Monoclonal specifically detects CD8 in Feline samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD8 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD8A | |
Feline CD8, Mr 70 kDa (38/31 kDa under reducing conditions). | |
0.1 mL | |
Adaptive Immunity, Cytokine Research, Immunology, Immunophenotyping, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
925 | |
Feline | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
fCD8 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
CD8, CD8a molecule, LEU2, p32 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Feline CD8, Mr 70 kDa (38/31 kDa under reducing conditions). | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction