Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
$489.50
Specifications
Antigen | CD8 |
---|---|
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1-2 ug/ml, Immunohistochemistry-Frozen |
Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoprecipitation |
Classification | Polyclonal |
Description
CD8 Polyclonal specifically detects CD8 in Human, Mouse, Porcine samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CD8 | |
Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1-2 ug/ml, Immunohistochemistry-Frozen | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Pig | |
P01732 | |
925 | |
Recombinant fragment from human CD8 C-terminal cytoplasmic domain of alpha chain (exact sequence is proprietary). (Uniprot: P01732) | |
Primary | |
Store at 4C. | |
32 kDa |
0.2 mg/ml | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
10 mM PBS, pH 7.4 with 0.05% BSA with 0.05% Sodium Azide | |
CD8, CD8a molecule, LEU2, p32 | |
CD8A | |
IgG | |
Protein A purified | |
CD8 is a cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha/beta) or as a homodimer (CD8 alpha/alpha). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title