Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (OX-8), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212523PECY55
Description
CD8 Monoclonal specifically detects CD8 in Rat samples. It is validated for Flow Cytometry.Specifications
CD8 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
CD8A | |
High Mw glycoproteins from rat thymocytes | |
0.1 mL | |
Primary | |
The mouse monoclonal antibody OX-8 recognizes the hinge-like membrane-proximal domain of rat CD8a (32-34 kDa; alpha chain of the CD8 antigen). | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OX-8 | |
Flow Cytometry | |
CD8, CD8a molecule, LEU2, p32 | |
Mouse | |
Protein A purified | |
Adaptive Immunity, Cytokine Research, Immunology, Immunophenotyping, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
925.0 | |
Rat | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction