Learn More
CD8 Antibody (RPA-T8), Alexa Fluor™ 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP225195AF350
Catalog No. NP2219A30
Only null left
Add to Cart
Description
Description
CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Flow (Cell Surface).
Specifications
Specifications
CD8
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
CD8A
CD8 (CD8a; alpha chain)
Primary
Human
IgG1 κ
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
RPA-T8
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Flow (Cell Surface)
CD8, CD8a molecule, LEU2, p32
Mouse
0.1 mL
925
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.