Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (RPA-T8), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
$511.50
Specifications
Antigen | CD8 |
---|---|
Clone | RPA-T8 |
Dilution | Flow Cytometry 1ul/1 million cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Flow Cytometry.Specifications
CD8 | |
Flow Cytometry 1ul/1 million cells | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD8, CD8a molecule, LEU2, p32 | |
CD8A | |
IgG1 κ | |
Protein G purified | |
This product reacts with Human. |
RPA-T8 | |
Flow Cytometry | |
APC | |
Mouse | |
Adaptive Immunity, Cytokine Research, Immunology, Phospho Specific, Signal Transduction, Stem Cell Markers | |
NP_741969 | |
925 | |
CD8 (CD8a; alpha chain) | |
Primary | |
Store at 4C. Do not freeze. | |
26 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title