Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (SP16), Novus Biologicals™

Rabbit Monoclonal Antibody
$243.50 - $890.00
Specifications
Antigen | CD8 |
---|---|
Clone | SP16 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CD8 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers | |
CD8, CD8a molecule, LEU2, p32 | |
CD8A | |
IgG | |
Tissue culture supernatant |
SP16 | |
Monoclonal | |
Supernatant | |
RUO | |
Human | |
925 | |
Synthetic peptide sequence comprising the 13 C-terminal amino acids of the cytoplasmic domain of the CD8 alpha chain. | |
Primary | |
Store at 4°C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title