Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Antibody (YTC182.20) - BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$192.94 - $532.07

Specifications

Antigen CD8
Clone YTC182.20
Concentration 1.0 mg/mL
Dilution Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Flow Cytometry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017665
SDP
View Documents
Novus Biologicals
NB100649220.025MG
0.025 mg
Each for $192.94
Only null left
Add to Cart
 
NB10064922
SDP
View Documents
Novus Biologicals
NB10064922
0.1 mg
Each for $532.07
Only null left
Add to Cart
 
Description

Description

CD8 Monoclonal specifically detects CD8 in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

CD8
1.0 mg/mL
Immunohistochemistry, Immunohistochemistry (Frozen), Flow Cytometry
Unconjugated
Rat
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers
P01732
925
Mouse L cells transfected with human CD8 gene
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
26 kDa
YTC182.20
Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Monoclonal
Purified
RUO
Human, Primate
CD8, CD8a molecule, LEU2, p32
CD8A
IgG2b
Protein G purified
Recognizes the human CD8 cell surface antigen, a 30/32kD glycoprotein expressed by the cytotoxic/suppressor subset of T lymphocytes, and more weakly by NK cells.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.