Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (YTC182.20), HRP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10064922H
Description
CD8 Monoclonal specifically detects CD8 in Human, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD8 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CD8A | |
Mouse L cells transfected with human CD8 gene | |
0.1 mL | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
Recognizes the human CD8 cell surface antigen, a 30/32kD glycoprotein expressed by the cytotoxic/suppressor subset of T lymphocytes, and more weakly by NK cells. | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunohistochemistry (Frozen) | |
YTC182.20 | |
Immunohistochemistry, Immunohistochemistry-Frozen | |
CD8, CD8a molecule, LEU2, p32 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human, Primate | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction