Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (YTS 105.18) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$281.00 - $599.00
Specifications
Antigen | CD8 alpha |
---|---|
Clone | YTS 105.18 |
Dilution | Flow Cytometry 1:10 - 1:1000, Immunohistochemistry-Frozen 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, Immunofluorescence 1:10 - 1:500, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry (Frozen), Blocking Assay, Immunofluorescence, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25265925
![]() |
Novus Biologicals
NBP2526590.025MG |
0.025 mg |
Each for $281.00
|
|
|||||
NBP25265902
![]() |
Novus Biologicals
NBP2526590.2MG |
0.2 mg |
Each for $599.00
|
|
|||||
Description
CD8 Monoclonal specifically detects CD8 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence, CyTOF-ready, Block/Neutralize.Specifications
CD8 alpha | |
Flow Cytometry 1:10 - 1:1000, Immunohistochemistry-Frozen 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, Immunofluorescence 1:10 - 1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD8, CD8 antigen, alpha polypeptide (p32), CD8a antigen, CD8a molecule, Leu2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, RPA-T8, RPA-T8 antibody flow, RPA-T8 CD8, RPA-T8 Clone, RPA-T8 Flow, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2 | |
CD8A | |
IgG | |
Protein A or G purified | |
Recognizes a non polymorphic epitope on the mouse CD8 alpha chain. This antibody has been reported to block MHC I dependent T cell responses in vitro and in vivo. |
YTS 105.18 | |
Flow Cytometry, Immunohistochemistry (Frozen), Blocking Assay, Immunofluorescence, CyTOF | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Cytokine Research, Immunology, Immunophenotyping, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
925 | |
Ly-2-transfected mouse L cells. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title