Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 beta Antibody (BU88), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2447530.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
CD8 beta Monoclonal antibody specifically detects CD8 beta in Human samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
CD8 beta | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain | |
Mouse | |
32 kDa | |
0.2 mg | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers | |
926 | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence | |
BU88 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CD8B | |
Human CD8 beta | |
Protein A or G purified | |
RUO | |
Primary | |
The T cell receptor (TCR) is a heterodimer composed of either alpha and beta or gamma and delta chains. CD3 chains and the CD4 or CD8 co-receptors are also required for efficient signal transduction through the TCR. The TCR is expressed on T helper and T cytotoxic cells that can be distinguished by their expression of CD4 and CD8. T helper cells express CD4 proteins and T cytotoxic cells display CD8. CD8 (also designated Leu 2 or T8), a cell surface glycoprotein, is a two chain complex (alpha-alpha or alpha-beta) receptor that binds class I MHC molecules presented by the antigen-presenting cell (APC). A primary function of CD8 is to facilitate antigen recognition by the TCR and to strengthen the avidity of the TCR-antigen interactions. An additional role for CD8-expressing T cells may be to maintain low levels of HIV expression. | |
Store at 4C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction