Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 beta Antibody (EP42), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128300PE
Description
CD8 beta Monoclonal specifically detects CD8 beta in Chicken samples. It is validated for Flow Cytometry.Specifications
CD8 beta | |
Monoclonal | |
PE | |
CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
beta chain (Mr 34 kDa) of chicken CD8 | |
Store at 4C in the dark. | |
IgG2a κ |
Flow Cytometry | |
EP42 | |
Flow Cytometry | |
CD8B | |
Chicken splenocytes | |
0.25 mL | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers | |
926 | |
Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction