Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 beta Antibody (H35-17.2), FITC, Novus Biologicals™

Rat Monoclonal Antibody
$622.00 - $780.00
Specifications
Antigen | CD8 beta |
---|---|
Clone | H35-17.2 |
Dilution | Flow Cytometry 1:10-1:1000 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14403801
![]() |
Novus Biologicals
NBP1440380.1MG |
0.1 mg |
Each for $622.00
|
|
|||||
NBP1440385
![]() |
Novus Biologicals
NBP1440380.5MG |
0.5 mg |
Each for $780.00
|
|
|||||
Description
CD8 beta Monoclonal specifically detects CD8 beta in Mouse samples. It is validated for Flow Cytometry.Specifications
CD8 beta | |
Flow Cytometry 1:10-1:1000 | |
Monoclonal | |
Rat | |
Innate Immunity, Stem Cell Markers | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
926 | |
The immunogen for this antibody was CD8. | |
Primary | |
Store at 4C. Do not freeze. |
H35-17.2 | |
Flow Cytometry | |
FITC | |
RUO | |
Mouse | |
CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain | |
CD8B | |
IgG2b κ | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title