Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 beta Antibody (H35-17.2), mFluor Violet 450 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP308222MFV450
Description
CD8 beta Monoclonal antibody specifically detects CD8 beta in Mouse, Human (Negative) samples. It is validated for Flow CytometrySpecifications
CD8 beta | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse, Human (Negative) | |
Purified |
Flow Cytometry | |
H35-17.2 | |
Flow Cytometry | |
CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain | |
Five day mixed leukocyte culture population, C57BL/6 anti-BALB/c (Uniprot: P10300) | |
0.1 mL | |
Adaptive Immunity, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers | |
926 | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction