Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Mouse anti-Human, APC, Clone: SK1, Proteintech
SDP

Catalog No. 502256695 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-6695 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-6695 Supplier Proteintech Group Inc Supplier No. APC65146100TESTS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The CD8 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD8. This antibody recognizes human antigen. The CD8 antibody has been validated for the following applications: FC analysis. This antibody is apc.

Specifications

Antigen CD8
Applications Flow Cytometry
Classification Monoclonal
Clone SK1
Concentration 5 μL (0.25 μg)/Test
Conjugate APC
Host Species Mouse
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 925
Target Species Human
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.