Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Mouse anti-Human, Baboon, Cynomolgus, Rhesus, BUV805, Clone: SK1, BD Horizon™

Catalog No. BDB612890 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612890 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612890 Supplier BD Biosciences Supplier No. 612890
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD8 Monoclonal antibody specifically detects CD8 in Human,Baboon,Cynomolgus,Rhesus samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8
Applications Flow Cytometry
Classification Monoclonal
Clone SK1
Conjugate Brilliant Ultraviolet 805
Host Species Mouse
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD8
Target Species Human, Baboon, Cynomolgus, Rhesus Monkey
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.