Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Mouse anti-Human, Brilliant Violet 786, Clone: G42-8, BD Optibuild™

Catalog No. BDB743069 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743069 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743069 Supplier BD Biosciences Supplier No. 743069
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The G42-8 monoclonal antibody specifically recognizes the alpha subunit (32 kDa) of CD8ααhomodimer or CD8αβheterodimer. The CD8 molecule binds to HLA class I molecules during interaction of CD8+ T cells with antigen-presenting cells or target cells. CD8 is expressed on T cytotoxic/suppressor cell populations. G42-8 stains approximately 13-48% of peripheral blood lymphocytes and 80% of thymocytes, as well as a subset of NK cells. This antibody is useful in detecting CD8+ T cells when using DimerX I reagents.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8
Classification Monoclonal
Clone G42-8
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD8α, CD8A, CD8 alpha, Leu2, MAL, T8, p32
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.