Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Mouse anti-Human, Clone: CA-8, Abnova™

Catalog No. 89933871 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-871 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-871 Supplier Abnova Corporation Supplier No. MAB12451
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length native of human CD8.

The CD8 antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. The CD8 antigen, acting as a coreceptor, and the T-cell receptor on the T lymphocyte recognize antigen displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. The functional coreceptor is either a homodimer composed of two alpha chains, or a heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. This gene encodes the CD8 beta chain isoforms. Multiple alternatively spliced transcript variants encoding distinct membrane associated or secreted isoforms have been described. A pseudogene, also located on chromosome 2, has been identified. [provided by RefSeq]

Specifications

Antigen CD8
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Clone CA-8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length native of human CD8.
Dilution Immunohistochemistry (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from ascites fluid (1.2% sodium acetate, 2mg BSA, 0.01mg sodium azide)
Gene Alias CD8/Leu2/MAL/p32
Gene Symbols CD8A
Host Species Mouse
Immunogen Native purified CD8 from human thymocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 925/926
Target Species Human
Content And Storage Store at −20°C. After reconstitution with 1mLof 1.2% sodium acetate or neutral PBS and concentration will be 100μg/mL, store at −20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.