Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Mouse, anti-Human, R718, Clone: G42-8, BD OptiBuild™

Catalog No. BDB751953 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751953 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751953 Supplier BD Biosciences Supplier No. 751953
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The G42-8 monoclonal antibody specifically recognizes the alpha subunit (32 kDa) of CD8αα homodimer or CD8αβ heterodimer. The CD8 molecule binds to HLA class I molecules during interaction of CD8+ T cells with antigen-presenting cells or target cells. CD8 is expressed on T cytotoxic/suppressor cell populations. G42-8 stains approximately 13-48% of peripheral blood lymphocytes and 80% of thymocytes, as well as a subset of NK cells. This antibody is useful in detecting CD8+ T cells when using DimerX I reagents.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8
Applications Flow Cytometry
Classification Monoclonal
Clone G42-8
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.