Learn More
CD8 Mouse anti-Human, Rhesus, Cynomolgus, Baboon, APC-R700, Clone: RPA-T8, BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD8 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | RPA-T8 |
| Conjugate | APC-R700 |
| Formulation | Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide. |
| Gene Alias | CD8α; CD8A; CD8 alpha; Leu2; MAL; T8; p32 |
| Host Species | Mouse |
| Immunogen | Human CD8a |
| Quantity | 100 Tests |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.