Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ CD8, Mouse, FITC, Clone: SK1,
Mouse Monoclonal Antibody
$284.50 - $2480.00
Specifications
Antigen | CD8 |
---|---|
Clone | SK1 |
Concentration | Please refer to lot specific datasheet. |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
MABF1670MI
|
MilliporeSigma™
MABF1670 |
FITC | 25 Tests |
Each for $284.50
|
|
||||
MABF1671MI
|
MilliporeSigma™
MABF1671 |
FITC | 100 Tests |
Each for $618.00
|
|
||||
MABF1672MI
|
MilliporeSigma™
MABF1672 |
FITC | 500 Tests |
Each for $2,480.00
|
|
||||
Description
The SK1 antibody is specific for the 32-34kDa alpha chain of human CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor for antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development, and cytotoxic effector functions. CD8+ cytotoxic T cells (CTLs) play an important role in inducing cell death of tumor cells, as well as cells infected by virus, bacteria or parasites.The SK1 antibody is widely used as a phenotypic marker for CD8 on cytotoxic T cells, thymocytes, as well as on certain cell types that do not also express the TCR, including some NK cells and lymphoid dendritic cells. It is cross-reactive with CD8 in several non-human species, including Baboon, Chimpanzee, Cynomolgus and Rhesus. If used together with an alternative Anti-Human CD8a clone, RPA-T8, the SK1 antibody will not block binding of RPA-T8 to CD8a.Specifications
CD8 | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P01732 | |
IgG1 κ | |
Purified by size-exclusion chromatography | |
CD8A, MAL |
SK1 | |
Flow Cytometry | |
FITC | |
Mouse | |
Inflammation & Immunology | |
Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2. | |
Immunogen information for this antibody is proprietary information. | |
Primary | |
Stable for 1 year at 2°-8°C from date of receipt. Note: It is recommended to store the product undiluted at 2°-8°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title