Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Rabbit anti-Mouse, Clone: 16, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | CD8 |
---|---|
Clone | 16 |
Dilution | Sandwich ELISA Capture 1:250-1:2000 |
Applications | ELISA |
Classification | Monoclonal |
Description
CD8 Monoclonal specifically detects CD8 in Mouse samples. It is validated for Sandwich ELISA Capture.Specifications
CD8 | |
Sandwich ELISA Capture 1:250-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD8, CD8a molecule, LEU2, p32 | |
Recombinant Mouse CD8 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
16 | |
ELISA | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
0.2 um filtered solution in PBS | |
925 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title