Learn More
CD80 (B7-1) Monoclonal Antibody (2D10.4), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$365.00
Specifications
Antigen | CD80 (B7-1) |
---|---|
Clone | 2D10.4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
17-080-942
![]() |
Invitrogen™
17080942 |
100 Tests |
Each of 1 for $365.00
|
![]() |
|||||
Description
Description: The 2D10.4 monoclonal antibody reacts with human CD80 (B7-1), a 60 kDa member of the Ig superfamily. CD80 is expressed by activated B cells, macrophages and dendritic cells. In addition, activated T cells express this antigen. CD80 has high affinity for binding to two T cell surface antigens, CD28 and CD152 (CTLA-4). The interaction of CD28 and CD152 with CD80 is crucial in T-B cell communication leading to activation of T and B cells, respectively. Applications Reported: This 2D10.4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2D10.4 antibody has been pre-diluted and tested by flow cytometric analysis of Daudi cell line. This may be used at 5 μL (1.0 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser
CD80 (B7-1) and CD86 (B7-2) are ligands of T cell critical costimulatory molecule CD28, and of an inhibitory receptor CTLA-4 (CD152). Both B7 molecules are expressed on professional antigen-presenting cells and are essential for T cell activation, and both molecules can also substitute for each other in this process. CD80 is rapidly induced on the surface of in vitro activated B cells, with Epstein Barr Virus (EBV) transformed B cell lines, with Burkitts lymphoma cell lines, with freshly isolated follicular B lymphoma cells, T cells, and monocytes. The B-lymphocyte activation antigen B7-1 provides regulatory signals for T lymphocytes as a consequence of binding to the CD28 and CTLA4 ligands of T cells. Diseases associated with CD80 dysfunction include gallbladder squamous cell carcinoma and myocarditis.Specifications
CD80 (B7-1) | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
941 | |
IgG1 κ | |
4°C, store in dark, DO NOT FREEZE! | |
CD80 |
2D10.4 | |
Monoclonal | |
Liquid | |
RUO | |
P33681 | |
CD80 | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.