Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD80 (B7-1) Mouse anti-Human, BUV737, Clone: 2D10.4 (also known as 2D10), BD OptiBuild™

Catalog No. BDB751732 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751732 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751732 Supplier BD Biosciences Supplier No. 751732
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD80 (B7-1) Monoclonal antibody specifically detects CD80 (B7-1) in Human samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD80 (B7-1)
Applications Flow Cytometry
Classification Monoclonal
Clone 2D10.4 (also known as 2D10)
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 737
Description Hu CD80 (B7-1) BUV737 2D10.4 50 μg
Host Species Mouse
Immunogen Not Reported
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD80 (B7-1)
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG1 κ
Reactivity Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.