Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD80/B7-1 Mouse anti-Human, Mouse, Porcine, Clone: 1E2F10, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | CD80/B7-1 |
---|---|
Clone | 1E2F10 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6834
![]() ![]() |
Proteintech Group Inc
664061IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Involved in the costimulatory signal essential for T lymphocytes activation. T cell proliferation and cytokine production is induced by the binding of CD28 or CTLA4 to this receptor.Specifications
CD80/B7-1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Activation B7 1 antigen, B7, B7 1, B7-1, BB1, CD28LG, CD28LG1, CD80, CD80 molecule, CTLA 4 counter receptor B7.1, LAB7 | |
CD80 | |
IgG1 | |
Protein G | |
Antibody |
1E2F10 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Pig | |
P33681, Q00609 | |
12519, 397161, 941 | |
CD80/B7-1 Fusion Protein Ag5615 | |
Primary | |
-20°C | |
CD80 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title