Learn More
CD80 (B7-1) Monoclonal Antibody (2D10.4), PerCP-eFluor™ 710, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
$244.00 - $492.00
Specifications
| Antigen | CD80 (B7-1) |
|---|---|
| Clone | 2D10.4 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
Description: The 2D10.4 monoclonal antibody reacts with human CD80 (B7-1), a 60 kDa member of the Ig superfamily. CD80 is expressed by activated B cells, macrophages and dendritic cells. In addition, activated T cells express this antigen. CD80 has high affinity for binding to two T cell surface antigens, CD28 and CD152 (CTLA-4). The interaction of CD28 and CD152 with CD80 is crucial in T-B cell communication leading to activation of T and B cells, respectively. Applications Reported: This 2D10.4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2D10.4 antibody has been pre-titrated and tested by flow cytometric analysis of Daudi cell line. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD80 (B7-1) and CD86 (B7-2) are ligands of T cell critical costimulatory molecule CD28, and of an inhibitory receptor CTLA-4 (CD152). Both B7 molecules are expressed on professional antigen-presenting cells and are essential for T cell activation, and both molecules can also substitute for each other in this process. CD80 is rapidly induced on the surface of in vitro activated B cells, with Epstein Barr Virus (EBV) transformed B cell lines, with Burkitts lymphoma cell lines, with freshly isolated follicular B lymphoma cells, T cells, and monocytes. The B-lymphocyte activation antigen B7-1 provides regulatory signals for T lymphocytes as a consequence of binding to the CD28 and CTLA4 ligands of T cells. Diseases associated with CD80 dysfunction include gallbladder squamous cell carcinoma and myocarditis.Specifications
| CD80 (B7-1) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Activation B7-1 antigen; B7; B7 protein; B7.1; B71; B7-1; B7-1 protein; B7-1 protein precursor; BB1; B-lymphocyte activation antigen B7; Cd28l; CD28LG; CD28LG1; CD80; Cd80 a rat homolog of the human CD28/CTLA - 4 ligand (B7-1); CD80 anitgen; CD80 antigen; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); Cd80 molecule; CD80 protein precursor; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; LAB7; Ly53; Ly-53; MIC17; sCD 80; sCD80; secreted B7-1 protein precursor; soluble CD 80; soluble CD80; T-cell costimulatory molecule; T-cell co-stimulatory protein B7-1; T-lymphocyte activation antigen CD80; TS/A-1; TSA1 | |
| CD80 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CD80 |
| 2D10.4 | |
| Flow Cytometry | |
| PerCP-eFluor 710 | |
| Mouse | |
| Human | |
| P33681 | |
| 941 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.