Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD80 (B7-1) Mouse, anti-Human, R718, Clone: 2D10.4, BD OptiBuild™

Catalog No. BDB751737 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751737 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751737 Supplier BD Biosciences Supplier No. 751737
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 2D10.4 monoclonal antibody, also known as 2D10, specifically recognizes CD80 which is otherwise known as Activation B7-1 antigen (B7-1), B7.1, or B7. This ∼60 kDa single-pass type I transmembrane glycoprotein is encoded by CD80 (CD80 molecule) that belongs to the immunoglobulin gene superfamily (IgSF). CD80 (B7-1) is comprised of an extracellular region with an IgV-like domain followed by an IgC-like domain, a transmembrane region and a cytoplasmic tail. It is variably expressed on activated B cells, activated CD4+ and CD8+ T cells, macrophages, and dendritic cells. CD80 (B7-1) serves as a ligand for the CD28 and CD152 (CTLA-4) coreceptors that are differentially expressed on T cells. CD80 (B7-1) engagement of CD28 costimulates T cells activated through their TCR-CD3 complex whereas its interaction with CD152 (CTLA-4) generates an inhibitory signal for T cell responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD80 (B7-1)
Applications Flow Cytometry
Classification Monoclonal
Clone 2D10.4
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.