Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD80/B7-1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501729345 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9345 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9345 Supplier Proteintech Group Inc Supplier No. 142921AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Involved in the costimulatory signal essential for T lymphocytes activation. T cell proliferation and cytokine production is induced by the binding of CD28 or CTLA4 to this receptor.

Specifications

Antigen CD80/B7-1
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CD80
Gene Accession No. P33681, Q00609
Gene Alias Activation B7 1 antigen, B7, B7 1, B7-1, BB1, CD28LG, CD28LG1, CD80, CD80 molecule, CTLA 4 counter receptor B7.1, LAB7
Gene Symbols CD80
Host Species Rabbit
Immunogen CD80/B7-1 Fusion Protein Ag5615
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12519, 941
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.