Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD80 Hamster anti-Mouse, FITC, Clone: 16-10A1, BD

Catalog No. BDB561954 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561954 0.1 mg
BDB553768 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561954 Supplier BD Biosciences Supplier No. 561954
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

Monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. Antibody was conjugated with FITC under optimum conditions, and unreacted FITC was removed. CD80 has been constitutively expressed on dendritic cells, monocytes, and peritoneal macrophages and it is inducible on B cells by various means.

Host Species: Hamster
Clone: 16-10A1
Isotype: IgG2, κ
Species Reactivity: Mouse
Immunogen: Mouse CD80 (B7) Transfected Cell Line

  • Activation of purified B cells with LPS can induce CD80 expression in as few as 18 hour
  • 16-10A1 antibody blocks binding of CTLA-4 Ig to CD80 and to block T-cell activation by Con A-elicited peritoneal exudate cells and CD80-transfected cell lines

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD80
Applications Flow Cytometry
Classification Monoclonal
Clone 16-10A1
Concentration 0.5mg/mL
Conjugate FITC
Description Cd80; B7; B7-1; Cd28l; Ly-53; MIC17; TSA1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Mouse CD80 (B7) Transfected Cell Line
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.