Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD80 Hamster anti-Mouse, PE, Clone: 16-10A1, BD

Catalog No. BDB561955 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561955 25 μg
BDB553769 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561955 Supplier BD Biosciences Supplier No. 561955
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The 16-10A1 antibody reacts with CD80 (B7-1). This member of the Ig superfamily, along with CD86 (B7-2), participates in T-cell co-stimulation via interactions with CD28 and CD152 (CTLA-4). CD80 has been reported to be constitutively expressed on dendritic cells, monocytes, and peritoneal macrophages; and it is inducible on B cells by various means, including activation by LPS, IL-4, and the cross-linking of surface Ig. Expression of CD80 has been reported to be greatly enhanced on splenic B cells following activation by LPS, with peak expression occurring between 48 and 72 hours. It has been reported that the activation of purified B cells with LPS can induce CD80 expression in as few as 18 hours. The 16-10A1 antibody has been reported to block binding of CTLA-4 Ig to CD80 and to block T-cell activation by Con A-elicited peritoneal exudate cells and CD80-transfected cell lines. However, 16-10A1 antibody alone is not able to block T-cell activation by antigen-presenting cells. CD86 (B7-2) is an alternate ligand for CD28 and CD152 (CTLA-4). Preliminary reports indicate that the 16-10A1 mAb may block the binding of rat anti-CD80 mAb clone 1G10 (Cat. No. 553368). In addition, it has been reported that the 16-10A1 antibody may cross-react with an activation antigen expressed on IFN-γ-activated alveolar macrophages of the dog.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD80
Applications Flow Cytometry
Classification Monoclonal
Clone 16-10A1
Concentration 0.2mg/mL
Conjugate PE
Description Cd80; B7; B7-1; Cd28l; Ly-53; MIC17; TSA1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Mouse CD80 (B7) Transfected Cell Line
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.