Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD81 Antibody (1.3.3.22), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$257.16 - $554.51

Specifications

Antigen CD81
Clone 1.3.3.22
Concentration 0.2 mg/mL
Dilution Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25331820
SDP
View Documents
Novus Biologicals
NBP25331820UG
20 μg
Each for $257.16
Only null left
Add to Cart
 
NB253318100
SDP
View Documents
Novus Biologicals
NBP253318100UG
100 μg
Each for $554.51
Only null left
Add to Cart
 
NB253318200
SDP
View Documents
Novus Biologicals
NBP253318200UG
200 μg N/A N/A N/A
Description

Description

CD81 Monoclonal specifically detects CD81 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Functional, Flow (Cell Surface), Knockout Validated, Single Cell Western.
Specifications

Specifications

CD81
0.2 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Adaptive Immunity, Cell Cycle and Replication, Immunology
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
975
B-Cell line derived from a Burkitt lymphoma
Primary
Store at 4C.
1.3.3.22
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Monoclonal
Purified
RUO
Human, Mouse, Rat
CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7
CD81
IgG1 κ
Protein A or G purified
26 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.