Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD81 Antibody (1D6), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281096PE
Description
CD81 Monoclonal specifically detects CD81 in Human, Goat, Chimpanzee, Sheep samples. It is validated for Flow Cytometry.Specifications
CD81 | |
Monoclonal | |
PE | |
PBS | |
CD81 | |
This antibody was produced by immunizing mice with 5A6-aggregated OCI-LY8 B cells. | |
0.1 mL | |
Adaptive Immunity, Cell Cycle and Replication, Immunology | |
975 | |
Human, Goat, Chimpanzee, Sheep | |
Purified |
Flow Cytometry | |
1D6 | |
Flow Cytometry | |
CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
This antibody is specific for human CD81, which is expressed on most tissues with the exception of red blood cells and platelets. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction