Learn More
CD81 Antibody (2F7), DyLight 405, Novus Biologicals™
SDP
Armenian Hamster Monoclonal Antibody
Supplier:  Novus Biologicals NBP128136V
Catalog No. NBP128136V
Only null left
Add to Cart
Description
Description
CD81 Monoclonal specifically detects CD81 in Human, Mouse, Hamster samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
CD81
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
CD81
Mouse epithelial cell line PAM212
0.25 mL
Adaptive Immunity, Cell Cycle and Replication, Immunology
975
Human, Mouse, Hamster
Purified
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
2F7
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7
Armenian Hamster
Protein A or G purified
RUO
Primary
Mouse CD81 (Mr 25 kDa), a member of the transmembrane 4 integral membrane protein family
Store at 4C in the dark.
IgG3 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.