Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD81 Antibody (C81/2885R), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308796R
Description
CD81 Monoclonal antibody specifically detects CD81 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry (Paraffin)Specifications
CD81 | |
Monoclonal | |
DyLight 550 | |
CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
Recombinant full-length human CD81 protein | |
100 μg | |
Adaptive Immunity, Cell Cycle and Replication, Immunology | |
975 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) | |
C81/2885R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction