Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD81 Antibody (M38), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP144861APCCY7
Description
CD81 Monoclonal specifically detects CD81 in Human, Mouse, Feline, Rabbit samples. It is validated for Flow Cytometry, Knockout Validated.Specifications
CD81 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
CD81 | |
MOLT-4, human T-ALL cell line (NM_004356.3). | |
0.1 mL | |
Primary | |
The antibody M38 reacts with CD81, a 25 kDa member of the tetraspanin family, expressed on majority of cells. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry, Immunoassay | |
M38 | |
Flow Cytometry, Knockout Validated | |
CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
Mouse | |
Protein A purified | |
Adaptive Immunity, Cell Cycle and Replication, Immunology | |
975.0 | |
Human, Feline, Rabbit | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction