Learn More
Invitrogen™ CD81 Monoclonal Antibody (2F7), APC

Description
CD81 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | CD81 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2F7 |
| Concentration | 0.1 mg/mL |
| Conjugate | APC |
| Formulation | PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.5 |
| Gene | CD81 |
| Gene Accession No. | P35762 |
| Gene Alias | 26 kDa cell surface protein TAPA-1; CD 81 antigen; Cd81; CD81 antigen; CD81 antigen (target of antiproliferative antibody 1); CD81 molecule; CVID6; S5.7; Tapa1; Tapa-1; Target of the antiproliferative antibody 1; Tetraspanin28; tetraspanin-28; Tspan28; Tspan-28 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.