Learn More
CD81 Mouse anti-Human, PE, Clone: JS-81, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555676
Description
JS-81 identifies the newly clustered CD81 antigen, a 26kDa, type III transmembrane protein (also known as TAPA-1) that is involved in cell growth and signal transduction. CD81 has a very broad cellular distribution, being expressed on cells of hematopoietic, neuroectodermal and mesenchymal origin. MAbs to CD81 have been shown to have anti-proliferative effects on different lymphoid cell lines, particularly those derived from large cell lymphomas. Immunoprecipitation studies reveal that CD81 is a component of a multimolecular complex which can include the Leu13 molecule, and the CD19 and CD21 molecules in B cells. MAbs to CD81 are reported to induce homotypic cell aggregation.
Host Species: Mouse
Clone: JS-81
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry

Specifications
CD81 | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
JS-81 | |
TAPA-1 | |
Mouse | |
100 Tests | |
Immunology | |
Canine, Human, Rabbit | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.