Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD81 Antibody (1D6), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281096FR
Description
CD81 Monoclonal specifically detects CD81 in Human, Goat, Chimpanzee, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CD81 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
CD81 | |
This antibody was produced by immunizing mice with 5A6-aggregated OCI-LY8 B cells. | |
0.1 mL | |
Adaptive Immunity, Cell Cycle and Replication, Immunology | |
975 | |
Human, Goat, Chimpanzee, Sheep | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1D6 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
This antibody is specific for human CD81, which is expressed on most tissues with the exception of red blood cells and platelets. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction