Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD81 Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: C81/2885R, Abnova™

Catalog No. 89339721 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-721 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-721 Supplier Abnova Corporation Supplier No. RAB00682
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CD81 Monoclonal antibody specifically detects CD81 in Human, Mouse, Rat samples. It is validated for ELISA, IHC-P.

Specifications

Antigen CD81
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C81/2885R
Conjugate Unconjugated
Description CD81 molecule
Gene CD81
Gene Alias S5.7, TAPA1, TSPAN28
Host Species Rabbit
Immunogen Recombinant protein corresponding to full-length human CD81
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 975
Target Species Human, Mouse, Rat
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.