Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CD81 Recombinant Protein
SDP

Catalog No. NB60215250U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB60215250U 250 μg
NB6021550UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB60215250U Supplier Novus Biologicals™ Supplier No. NBP260215250UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids 113-201 of Human CD81 The Recombinant Human CD81 Protein is derived from Baculovirus. The Recombinant Human CD81 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4) containing 10% glycerol. containingno Preservative
Gene ID (Entrez) 975
Name Recombinant CD81 Protein
Purification Method >95%, by SDS-PAGE
Quantity 250 μg
Source Baculovirus
Storage Requirements Store at 4°Cshort term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1ug of protein (determined by LAL method)
Gene Alias CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7
Gene Symbol CD81
Product Type Recombinant Protein
Recombinant Recombinant
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.