Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD81 (TAPA-1) Mouse, anti-Human, R718, Clone: JS-81, BD OptiBuild™

Catalog No. BDB751941 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751941 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751941 Supplier BD Biosciences Supplier No. 751941
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The JS-81 monoclonal antibody specifically binds to CD81, which is also known as, Target of the antiproliferative antibody 1 (TAPA1, TAPA-1), or Tetraspanin-28 (Tspan-28/TSPAN28). CD81 is an ∼26 kDa transmembrane protein that belongs to the tetraspanin (TM4SF) family. It is involved in cell growth and signal transduction. CD81 has a very broad cellular distribution, being expressed on cells of hematopoietic, neuroectodermal and mesenchymal origin. In hematopoietic cells, the CD81 antigen is expressed on B and T lymphocytes, NK cells, thymocytes, eosinophils, germinal center follicular dendritic cells, and to a variable extent on monocytes. The CD81 antigen is not expressed on neutrophils, platelets, or erythrocytes. CD81-specific antibodies have been shown to have anti-proliferative effects on different lymphoid cell lines, particularly those derived from large cell lymphomas. They are also reported to induce homotypic cell aggregation. Immunoprecipitation studies reveal that CD81 is a component of a multimolecular complex of CD19, CD21, and CD225 that is involved in the activation and control of B cell growth.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD81 (TAPA-1)
Applications Flow Cytometry
Classification Monoclonal
Clone JS-81
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.