Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD82/Kai-1 Antibody (C33), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221792APCCY7
Description
CD82/Kai-1 Monoclonal specifically detects CD82/Kai-1 in Human samples. It is validated for Flow Cytometry.Specifications
CD82/Kai-1 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
CD82 | |
C91/PL (human HTLV-1+ T cell line). | |
0.1 mL | |
Primary | |
The antibody C33 reacts with CD82, a widely expressed cell surface protein of the tetraspanin family. CD82 is also found in endosome/lysosome compartments. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
C33 | |
Flow Cytometry | |
CD82 antigenC33, CD82 molecule, IA4C33 antigen, Inducible membrane protein R2, KAI1GR15, kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 leukocyteantigen, antigen detected by monoclonal and IA4)), Metastasis suppressor Kangai-1, R2, SAR2tetraspanin-27, ST6tspan-27, Suppressor of tumorigenicity 6 protein, Tetraspanin-27, Tspan-27, TSPAN274F9 | |
Mouse | |
Protein A purified | |
Cancer | |
3732.0 | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction