Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD82 Mouse anti-Human, Mouse, Clone: 1E7A4, Proteintech
SDP

Catalog No. 501737201 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7201 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7201 Supplier Proteintech Group Inc Supplier No. 668031IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This metastasis suppressor gene product is a membrane glycoprotein that is a member of the transmembrane 4 superfamily. Expression of this gene has been shown to be downregulated in tumor progression of human cancers and can be activated by p53 through a consensus binding sequence in the promoter. Its expression and that of p53 are strongly correlated, and the loss of expression of these two proteins is associated with poor survival for prostate cancer patients. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

Specifications

Antigen CD82
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1E7A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CD82
Gene Accession No. P27701, P40237
Gene Alias 4F9, C33, C33 antigen, CD82, CD82 antigen, CD82 molecule, GR15, IA4, Inducible membrane protein R2, KAI1, Metastasis suppressor Kangai 1, R2, SAR2, ST6, Tetraspanin 27, Tspan 27, TSPAN27
Gene Symbols CD82
Host Species Mouse
Immunogen CD82 Fusion Protein Ag17513
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12521, 3732
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.