Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD83 Antibody (004), HRP

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290917H
Description
CD83 Monoclonal specifically detects CD83 in Rat samples. It is validated for ELISA.Specifications
CD83 | |
Monoclonal | |
HRP | |
B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD83 (B2GV95; Met1-Ala134). | |
0.1 mL | |
Dendritic Cell Markers | |
9308 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
4 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction