Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD83 Antibody (017), Janelia Fluor™ 646 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289979JF646
Description
CD83 Monoclonal antibody specifically detects CD83 in Human samples. It is validated for ELISASpecifications
| CD83 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CD83 (Q01151-1; Met 1-Ala 143). | |
| 0.1 mL | |
| Dendritic Cell Markers | |
| 9308 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 017 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction