Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD83 Antibody (HB15e), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1774W
This item is not returnable.
View return policy
Description
CD83 Monoclonal antibody specifically detects CD83 in Human samples. It is validated for Flow CytometrySpecifications
| CD83 | |
| Monoclonal | |
| DyLight 650 | |
| B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83 | |
| COS-7 African green monkey SV40 transformed kidney fibroblast-like cells transfected with human CD83 | |
| 0.5 mL | |
| Dendritic Cell Markers | |
| 9308 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| HB15e | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction