Learn More
Invitrogen™ CD83 Recombinant Rabbit Monoclonal Antibody (105), FITC

Description
Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human CD83 (Met1-Ala143) and conjugated with FITC under optimum conditions, the unreacted FITC was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.
Specifications
Specifications
| Antigen | CD83 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | 105 |
| Concentration | 100 μg/mL |
| Conjugate | FITC |
| Formulation | PBS with 0.5% BSA and 0.09% sodium azide |
| Gene | CD83 |
| Gene Accession No. | Q01151 |
| Gene Alias | B-cell activation protein; BL11; CD antigen CD83; CD83; CD83 antigen; CD83 antigen (activated B lymphocytes, immunoglobulin superfamily); CD83 molecule; cell surface protein HB15; cell-surface glycoprotein; HB15; hCD83; mCD83 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.