Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD84 Monoclonal Antibody (05)
CD84 Monoclonal Antibody (5)
Supplier: Thermo Scientific MA530194
Description

Specifications
CD84 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9UIB8 | |
CD84 | |
Recombinant Human CD84 Protein (Met1-Gly225). | |
100 μL | |
Primary | |
Human | |
Antibody |
ELISA | |
05 | |
Unconjugated | |
CD84 | |
A130013D22Rik; CD84; CD84 antigen; CD84 antigen (leukocyte antigen); CD84 molecule; CDw84; cell surface antigen MAX.3; DKFZp781E2378; hCD84; Hly9-beta; leucocyte differentiation antigen CD84; leukocyte antigen CD84; leukocyte differentiation antigen CD84; LY9B; mCD84; RGD1564553; Signaling lymphocytic activation molecule 5; SLAM family member 5; SLAMF5 | |
Mouse | |
Protein A | |
RUO | |
8832 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction